Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011528.4 | NP_035658.1 | 8 | 1075 | ||
GenBank mRNA | AK003092.1 | - | 8 | 1039 | ||
AK142030.1 | - | 8 | 1033 | |||
AK198205.1 | - | 1 | 44 | |||
BC003970.1 | - | 8 | 1008 | |||
BC004754.1 | - | 8 | 1030 | 3584511 | ||
BC094277.1 | - | 8 | 1024 | 5008399 | ||
U67611.1 | - | 8 | 1018 |