Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011536.2 | NP_035666.2 | 3 | 355 | ||
NM_172798.1 | NP_766386.1 | 2 | 305 | |||
GenBank mRNA | AB221665.1 | - | 3 | 355 | ||
AK028407.1 | - | 2 | 239 | |||
AK139949.1 | - | 2 | 277 | |||
AK147984.1 | - | 2 | 223 | |||
AY075134.1 | - | 3 | 355 | |||
BQ560189.1 | - | 2 | 305 | |||
U57329.1 | - | 2 | 82 |