Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159384.1 | NP_001152856.1 | 4 | 272 | ||
NM_001159385.1 | NP_001152857.1 | 5 | 362 | |||
NM_011550.3 | NP_035680.3 | 5 | 524 | |||
GenBank mRNA | AF213670.1 | - | 4 | 236 | ||
AF213671.1 | - | 5 | 326 | |||
AF213672.1 | - | 5 | 488 | |||
AK011478.1 | - | 4 | 252 | |||
AK142605.1 | - | 5 | 328 | |||
BC031380.1 | - | 5 | 504 | 5252695 | ||
BC038004.1 | - | 5 | 504 | |||
BC080768.1 | - | 4 | 1329 | |||
U43548.1 | - | 5 | 484 | |||
U43607.1 | - | 5 | 495 |