Search Thermo Fisher Scientific
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100169.1 | NP_001093639.1 | 13 | 1902 | ||
NM_001100170.1 | NP_001093640.1 | 13 | 1872 | |||
NM_006839.2 | NP_006830.2 | 13 | 1905 | |||
XM_005264110.3 | XP_005264167.1 | 13 | 1541 | |||
XM_005264113.1 | XP_005264170.1 | 12 | 1477 | |||
XM_005264114.1 | XP_005264171.1 | 12 | 1448 | |||
XM_011532497.2 | XP_011530799.1 | 13 | 1565 | |||
XM_017003194.1 | XP_016858683.1 | 13 | 1573 | |||
XM_017003195.1 | XP_016858684.1 | 13 | 1597 | |||
GenBank mRNA | AK225780.1 | - | 13 | 1539 | ||
AK296676.1 | - | 13 | 1511 | |||
AK299014.1 | - | 12 | 1427 | |||
AK299596.1 | - | 11 | 1282 | |||
AK300017.1 | - | 11 | 1248 | |||
AK312251.1 | - | 13 | 1555 | |||
AY232292.1 | - | 13 | 1535 | |||
AY921637.1 | - | 13 | 1553 | |||
BC002412.2 | - | 13 | 1546 | 2964676 | ||
BC027458.1 | - | 2 | 258 | |||
BX537369.1 | - | 13 | 1576 | |||
CR456793.1 | - | 13 | 1517 | |||
CR749590.1 | - | 8 | 949 | |||
D21092.1 | - | 11 | 1407 | |||
D21093.1 | - | 2 | 169 | |||
D21094.1 | - | 13 | 1602 | |||
L42572.1 | - | 13 | 1609 |