Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039363.2 | NP_001034452.1 | 12 | 1339 | ||
NM_001286713.1 | NP_001273642.1 | 12 | 1339 | |||
NM_001286714.1 | NP_001273643.1 | 12 | 1334 | |||
NM_001286716.1 | NP_001273645.1 | 12 | 1334 | |||
NM_001286718.1 | NP_001273647.1 | 12 | 1339 | |||
NM_001286720.1 | NP_001273649.1 | 12 | 1339 | |||
NM_011647.3 | NP_035777.2 | 12 | 1339 | |||
GenBank mRNA | AF132986.1 | - | 8 | 867 | ||
AK143799.1 | - | 12 | 1340 | |||
AK147016.1 | - | 12 | 1315 | |||
AK147302.1 | - | 12 | 1279 | |||
AK147487.1 | - | 12 | 1308 | |||
AK147874.1 | - | 12 | 1304 | |||
AK147908.1 | - | 12 | 1340 | |||
AK148062.1 | - | 12 | 1331 | |||
AK148072.1 | - | 12 | 1236 | |||
AK170589.1 | - | 12 | 1330 | |||
AK171007.1 | - | 12 | 1308 | |||
AK217801.1 | - | 1 | 88 | |||
AK218975.1 | - | 1 | 88 | |||
BC052449.1 | - | 12 | 1315 | 6409037 | ||
BC060701.1 | - | 12 | 1315 | 6409037 | ||
U37775.1 | - | 11 | 1251 | |||
U39818.1 | - | 12 | 1310 |