Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177609.1 | NP_001171080.1 | 8 | 1289 | ||
NM_001177610.1 | NP_001171081.1 | 7 | 578 | |||
NM_011665.4 | NP_035795.1 | 7 | 616 | |||
GenBank mRNA | AK003141.1 | - | 7 | 523 | ||
AK005058.1 | - | 7 | 598 | |||
AK011239.1 | - | 7 | 524 | |||
AK012282.1 | - | 7 | 542 | |||
AK088508.1 | - | 7 | 601 | |||
AK150575.1 | - | 7 | 559 | |||
AK160014.1 | - | 7 | 545 | |||
AK160988.1 | - | 7 | 542 | |||
AK165606.1 | - | 7 | 528 | |||
AK165615.1 | - | 7 | 546 | |||
AK166016.1 | - | 7 | 526 | |||
AK166657.1 | - | 8 | 1290 | |||
AK166930.1 | - | 7 | 522 | |||
AK167162.1 | - | 8 | 759 | |||
AK168212.1 | - | 7 | 545 | |||
AK168706.1 | - | 7 | 505 | |||
AK168766.1 | - | 7 | 539 | |||
U31934.1 | - | 6 | 456 | |||
U76416.1 | - | 7 | 552 | |||
U82627.1 | - | 7 | 561 | |||
U94402.1 | - | 7 | 612 | |||
X97575.1 | - | 7 | 493 | |||
X99739.1 | - | 7 | 540 |