Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177609.1 | NP_001171080.1 | 6, 7 | 1164 | ||
NM_001177610.1 | NP_001171081.1 | 5, 6 | 453 | |||
NM_011665.4 | NP_035795.1 | 5, 6 | 491 | |||
GenBank mRNA | AK003141.1 | - | 5, 6 | 398 | ||
AK005058.1 | - | 5, 6 | 473 | |||
AK011239.1 | - | 5, 6 | 399 | |||
AK012282.1 | - | 5, 6 | 417 | |||
AK088508.1 | - | 5, 6 | 476 | |||
AK150575.1 | - | 5, 6 | 434 | |||
AK160014.1 | - | 5, 6 | 420 | |||
AK160988.1 | - | 5, 6 | 417 | |||
AK165606.1 | - | 5, 6 | 403 | |||
AK165615.1 | - | 5, 6 | 421 | |||
AK166016.1 | - | 5, 6 | 401 | |||
AK166657.1 | - | 6, 7 | 1165 | |||
AK166930.1 | - | 5, 6 | 397 | |||
AK167162.1 | - | 6, 7 | 634 | |||
AK168212.1 | - | 5, 6 | 420 | |||
AK168706.1 | - | 5, 6 | 380 | |||
AK168766.1 | - | 5, 6 | 414 | |||
U31934.1 | - | 4, 5 | 331 | |||
U76416.1 | - | 5, 6 | 427 | |||
U82627.1 | - | 5, 6 | 436 | |||
U94402.1 | - | 5, 6 | 487 | |||
X97575.1 | - | 5, 6 | 368 | |||
X99739.1 | - | 5, 6 | 415 |