Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011704.3 | NP_035834.2 | 4 | 831 | ||
GenBank mRNA | AJ132098.1 | - | 4 | 664 | ||
AK002773.1 | - | 4 | 663 | |||
AK145984.1 | - | 4 | 737 | |||
AK149485.1 | - | 2 | 212 | |||
AK167427.1 | - | 4 | 663 | |||
BC019203.1 | - | 4 | 657 | 5041670 |