Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130169.1 | NP_001123641.1 | 4 | 589 | ||
NM_001130170.1 | NP_001123642.1 | 4 | 589 | |||
NM_001130171.1 | NP_001123643.1 | 4 | 589 | |||
NM_011751.3 | NP_035881.1 | 4 | 589 | |||
GenBank mRNA | AB013357.1 | - | 4 | 523 | ||
AB032196.1 | - | 4 | 470 | |||
AK033497.1 | - | 4 | 538 | |||
AK087885.1 | - | 4 | 589 | |||
AK140139.1 | - | 4 | 573 | |||
AK146787.1 | - | 4 | 552 | |||
AK159683.1 | - | 4 | 540 | |||
BC003715.1 | - | 4 | 530 | 3601357 |