Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EIF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201478.1 | NP_001188407.1 | 2 | 352 | ||
NM_006893.2 | NP_008824.2 | 2 | 352 | |||
XM_005277509.3 | XP_005277566.1 | 2 | 267 | |||
XM_006711196.3 | XP_006711259.1 | 2 | 267 | |||
XM_011509257.2 | XP_011507559.1 | 2 | 267 | |||
XM_011509258.2 | XP_011507560.1 | 2 | 267 | |||
XM_017000530.1 | XP_016856019.1 | 2 | 267 | |||
XM_017000531.1 | XP_016856020.1 | 2 | 267 | |||
XM_017000532.1 | XP_016856021.1 | 2 | 267 | |||
GenBank mRNA | AF220417.2 | - | 2 | 260 | ||
AF262403.1 | - | 2 | 260 | |||
AK001852.1 | - | 2 | 265 | |||
AK225259.1 | - | 2 | 246 | |||
AK294536.1 | - | 2 | 266 | |||
BC001585.1 | - | 2 | 250 | 3463026 | ||
BC039134.1 | - | 2 | 203 | 4339127 | ||
BC058905.1 | - | 2 | 217 | 6067337 | ||
BI465059.1 | - | 2 | 357 | |||
DC399556.1 | - | 2 | 352 |