Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110328.1 | NP_001103798.1 | 3 | 1498 | ||
NM_001110329.1 | NP_001103799.1 | 3 | 1441 | |||
NM_001110330.1 | NP_001103800.1 | 3 | 1371 | |||
NM_011813.3 | NP_035943.3 | 3 | 1482 | |||
GenBank mRNA | AF126746.1 | - | 3 | 1432 | ||
AF126747.1 | - | 3 | 1440 | |||
AK003641.1 | - | 1 | 592 | |||
AK135672.1 | - | 3 | 1372 | |||
AK142205.1 | - | 3 | 1391 | |||
AK162454.1 | - | 3 | 1363 | |||
AK170633.1 | - | 3 | 1367 | |||
AK220212.1 | - | 3 | 1371 | |||
BC006633.1 | - | 3 | 1482 | 3482191 | ||
BC106113.1 | - | 3 | 1357 | 6492060 | ||
BC138231.1 | - | 3 | 1432 | 8861250 | ||
BC138232.1 | - | 3 | 1457 | 8861251 | ||
BC145207.1 | - | 3 | 1457 | 9053758 |