Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011835.2 | NP_035965.2 | 11 | 1525 | ||
GenBank mRNA | AF153197.1 | - | 11 | 1503 | ||
AK012319.1 | - | 11 | 1523 | |||
AK041900.1 | - | 11 | 1519 | |||
AV135581.1 | - | 1 | 21 | |||
BC009136.1 | - | 11 | 1439 | 3597742 |