Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162947.1 | NP_001156419.1 | 7, 8 | 653 | ||
NM_011848.4 | NP_035978.2 | 8, 9 | 711 | |||
GenBank mRNA | AF093416.1 | - | 7, 8 | 658 | ||
AF099066.1 | - | 8, 9 | 680 | |||
BC005411.1 | - | 8, 9 | 688 | 3590866 | ||
CV560683.1 | - | 1, 2 | 65 |