Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011873.2 | NP_036003.2 | 2 | 158 | ||
GenBank mRNA | AF085348.1 | - | 2 | 123 | ||
AK002595.1 | - | 2 | 195 | |||
AK050159.1 | - | 2 | 172 | |||
AK088916.1 | - | 2 | 180 | |||
AK132139.1 | - | 2 | 172 | |||
AK137246.1 | - | 2 | 159 | |||
AK145398.1 | - | 2 | 192 | |||
AK146970.1 | - | 2 | 183 | |||
AK151730.1 | - | 1 | 874 | |||
AK152511.1 | - | 1 | 874 | |||
AK169769.1 | - | 1 | 458 | |||
BC014808.1 | - | 2 | 158 | 4506124 | ||
BC029351.1 | - | 1 | 89 | |||
BC083347.1 | - | 2 | 150 | 6829350 |