Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011886.2 | NP_036016.2 | 9 | 1041 | ||
GenBank mRNA | AF005036.1 | - | 9 | 948 | ||
AF295403.1 | - | 9 | 946 | |||
AK135335.1 | - | 9 | 1025 | |||
AK146204.1 | - | 9 | 1043 | |||
AK160549.1 | - | 9 | 1026 | |||
AK170115.1 | - | 8 | 906 | |||
BC002021.1 | - | 9 | 1039 | 3489293 |