Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011886.2 | NP_036016.2 | 9 | 1213 | ||
GenBank mRNA | AF005036.1 | - | 9 | 1120 | ||
AK135335.1 | - | 9 | 1197 | |||
AK146204.1 | - | 9 | 1215 | |||
AK160549.1 | - | 9 | 1198 | |||
AK170115.1 | - | 8 | 1078 | |||
AK186909.1 | - | 1 | 43 | |||
BC002021.1 | - | 9 | 1211 | 3489293 |