Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF176903.1 | - | 3 | 2045 | ||
AK014578.1 | - | 3 | 2063 | |||
AK140405.1 | - | 3 | 2057 | |||
BC039139.1 | - | 4 | 1904 | 3672353 | ||
BC051977.1 | - | 2 | 2350 | |||
BC053428.1 | - | 3 | 2050 | 6506766 | ||
BC069914.1 | - | 1 | 1567 | 5251876 |