Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Srp54a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011899.4 | NP_036029.2 | 7, 8 | 766 | ||
GenBank mRNA | AK011928.1 | - | 7, 8 | 730 | ||
BC019683.1 | - | 7, 8 | 695 | 3708250 |
Gene Symbol: Srp54b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100109.1 | NP_001093579.1 | 7, 8 | 1002 | ||
GenBank mRNA | AK136033.1 | - | 7, 8 | 1003 | ||
X16319.1 | - | 731 |