Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305033.1 | NP_001291962.1 | 4 | 1176 | ||
NM_001305035.1 | NP_001291964.1 | 6 | 1306 | |||
NM_001305036.1 | NP_001291965.1 | 5 | 1234 | |||
NM_001305037.1 | NP_001291966.1 | 5 | 1161 | |||
NM_001305038.1 | NP_001291967.1 | 5 | 1253 | |||
NM_001305039.1 | NP_001291968.1 | 6 | 1311 | |||
NM_001305040.1 | NP_001291969.1 | 7 | 1387 | |||
NM_006955.2 | NP_008886.1 | 5 | 1229 | |||
GenBank mRNA | AJ491697.1 | - | 5 | 936 | ||
AK025638.1 | - | 1 | 310 | |||
AK304306.1 | - | 5 | 860 | |||
BC048313.2 | - | 5 | 934 | |||
BC065181.1 | - | 6 | 988 | |||
BC107726.1 | - | 5 | 927 | |||
X68684.1 | - | 2 | 596 |