Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011968.3 | NP_036098.1 | 3 | 317 | ||
GenBank mRNA | AF060087.1 | - | 3 | 226 | ||
AK088143.1 | - | 3 | 255 | |||
AK152915.1 | - | 3 | 276 | |||
AK161662.1 | - | 3 | 256 | |||
BC086685.1 | - | 3 | 244 | 30681804 | ||
U60288.1 | - | 3 | 259 |