Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006993.2 | NP_008924.1 | 2 | 190 | ||
GenBank mRNA | AF081280.1 | - | 2 | 178 | ||
AY049737.1 | - | 2 | 182 | |||
BC041067.1 | - | 2 | 171 | 5473410 | ||
BC054868.1 | - | 2 | 176 | 6496849 | ||
BM739536.1 | - | 2 | 190 | |||
BM765409.1 | - | 2 | 190 |