Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081457.2 | NP_001074926.1 | 12 | 1403 | ||
NM_001081458.2 | NP_001074927.1 | 12 | 1403 | |||
NM_001135001.1 | NP_001128473.1 | 14 | 1634 | |||
NM_012023.3 | NP_036153.2 | 12 | 1403 | |||
GenBank mRNA | AB055635.1 | - | 12 | 1253 | ||
AB055636.1 | - | 12 | 1253 | |||
AK031584.1 | - | 12 | 1406 | |||
AK047089.1 | - | 3 | 167 | |||
AK129042.1 | - | 13 | 2604 | |||
AK139212.1 | - | 1 | 1138 | |||
AK178752.1 | - | 3 | 161 | |||
AK211769.1 | - | 3 | 158 | |||
AY389498.1 | - | 4 | 308 | |||
BC003979.1 | - | 12 | 1363 | 3489999 | ||
CN532075.1 | - | 4 | 353 | |||
CX568604.1 | - | 4 | 327 | |||
U37353.1 | - | 12 | 1232 |