Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083959.1 | NP_001077428.1 | 3 | 326 | ||
NM_001083960.1 | NP_001077429.1 | 3 | 326 | |||
NM_012046.2 | NP_036176.1 | 4 | 440 | |||
GenBank mRNA | AF126400.1 | - | 3 | 326 | ||
AF149309.1 | - | 3 | 289 | |||
AF163053.1 | - | 3 | 326 | |||
AF163054.1 | - | 3 | 326 | |||
AF165313.1 | - | 3 | 319 | |||
AF167439.1 | - | 3 | 319 | |||
AF169386.1 | - | 4 | 437 | |||
AF173848.1 | - | 4 | 433 | |||
AK088498.1 | - | 3 | 643 |