Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242580.1 | NP_001229509.1 | 5 | 583 | ||
NM_012049.2 | NP_036179.1 | 5 | 551 | |||
GenBank mRNA | AF069988.1 | - | 5 | 532 | ||
AK002269.1 | - | 5 | 591 | |||
AK004988.2 | - | 544 | ||||
AK032189.1 | - | 1234 | ||||
BC021634.1 | - | 5 | 533 | 4008543 |