Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083967.1 | NP_001077436.1 | 20 | 3204 | ||
NM_013685.2 | NP_038713.1 | 20 | 3531 | |||
GenBank mRNA | AK017365.1 | - | 1 | 84 | ||
AK028748.1 | - | 20 | 3129 | |||
AK078780.1 | - | 3 | 1682 | |||
AK081147.1 | - | 1 | 811 | |||
AK089036.1 | - | 1 | 929 | |||
AK132140.1 | - | 13 | 2545 | |||
AK133885.1 | - | 21 | 3943 | |||
BC043050.1 | - | 19 | 2991 | 5708370 | ||
U16322.1 | - | 21 | 3484 |