Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Uncx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013702.3 | NP_038730.1 | 3 | 2014 | ||
GenBank mRNA | AF247550.1 | - | 3 | 1983 | ||
AJ001116.1 | - | 3 | 1954 | |||
AV120135.1 | - | 1 | 114 | |||
BC051973.1 | - | 3 | 1974 | 5718115 |