Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3yl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289480.1 | NP_001276409.1 | 6 | 998 | ||
NM_013709.5 | NP_038737.1 | 7 | 1112 | |||
GenBank mRNA | AK141199.1 | - | 7 | 1113 | ||
AK168483.1 | - | 8 | 738 | |||
BC010296.1 | - | 6 | 560 | 3154966 | ||
BC058557.1 | - | 6 | 975 | 6313231 | ||
BC061488.1 | - | 7 | 1052 | 4483634 | ||
BC110043.1 | - | 7 | 1090 | 5149633 | ||
D85926.1 | - | 7 | 595 |