Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048146.2 | NP_001041611.1 | 3 | 726 | ||
NM_001286507.1 | NP_001273436.1 | 3 | 726 | |||
NM_001286508.1 | NP_001273437.1 | 3 | 644 | |||
NM_013727.4 | NP_038755.1 | 3 | 726 | |||
GenBank mRNA | AB007141.1 | - | 3 | 562 | ||
AK004992.1 | - | 3 | 705 | |||
AK015942.1 | - | 3 | 547 | |||
AK030076.1 | - | 3 | 558 | |||
AK036102.1 | - | 3 | 633 | |||
AK134714.1 | - | 3 | 629 | |||
AK153477.1 | - | 3 | 690 | |||
AK165549.1 | - | 3 | 612 | |||
AK167783.1 | - | 3 | 670 | |||
BC006851.1 | - | 3 | 621 | 3156857 |