Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mixl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013729.3 | NP_038757.1 | 2 | 1498 | ||
GenBank mRNA | AF154573.1 | - | 2 | 1499 | ||
AF201959.1 | - | 2 | 1448 | |||
AK138043.1 | - | 1 | 813 |