Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013737.5 | NP_038765.2 | 5, 6 | 848 | ||
GenBank mRNA | AK005158.1 | - | 5, 6 | 585 | ||
AK005210.1 | - | 5, 6 | 642 | |||
AK051454.1 | - | 5, 6 | 849 | |||
AK155678.1 | - | 6, 7 | 699 | |||
BC010726.1 | - | 5, 6 | 604 | 3707891 | ||
U34277.1 | - | 5, 6 | 594 |