Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013841.3 | NP_038869.1 | 14 | 2189 | ||
GenBank mRNA | AK146190.1 | - | 14 | 1591 | ||
AK168101.1 | - | 14 | 1591 | |||
BC012691.1 | - | 14 | 1565 | 4038142 | ||
BC058528.1 | - | 14 | 1581 | 5718352 | ||
U66865.1 | - | 14 | 2204 |