Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hunk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015755.2 | NP_056570.1 | 11 | 2532 | ||
GenBank mRNA | AF055919.2 | - | 11 | 2532 | ||
AF167987.1 | - | 11 | 2540 | |||
AK144639.1 | - | 11 | 2515 | |||
AK147425.1 | - | 11 | 2918 | |||
AK147734.1 | - | 11 | 2286 | |||
BC137788.1 | - | 11 | 2589 | 8860807 | ||
BC137790.1 | - | 11 | 2588 | 8860809 |