Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006905.2 | NP_008836.2 | 5 | 1568 | ||
XM_005259065.3 | XP_005259122.1 | 4 | 1288 | |||
XM_011527125.1 | XP_011525427.1 | 4 | 1288 | |||
GenBank mRNA | BC022338.1 | - | 5 | 1568 | 4732357 | |
J04539.1 | - | 5 | 1490 | |||
M33663.1 | - | 5 | 1493 | |||
M34715.1 | - | 5 | 1591 | |||
M37397.1 | - | 5 | 1520 |