Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018822.3 | NP_061292.2 | 8 | 1231 | ||
GenBank mRNA | AF153827.1 | - | 8 | 1205 | ||
AF304053.1 | - | 8 | 1205 | |||
AF304054.1 | - | 8 | 1205 | |||
AK034220.1 | - | 8 | 1035 | |||
AK052266.1 | - | 8 | 1224 | |||
AK076267.1 | - | 8 | 1261 | |||
AK128926.1 | - | 8 | 1139 | |||
AK141266.1 | - | 8 | 1179 | |||
BC130268.1 | - | 8 | 1228 | 40143074 |