Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCN SIRNA Assays ›
Gene Symbol:
Ucn
Gene Name:
urocortin
Gene Aliases:
Mpv17, Ucn1
Chromosome Location:
Chr.5: 31137989 - 31138895 on Build GRCm38
UniGene ID:
Mm.377116
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
188774
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ucn
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021290.2
NP_067265.1
2
603
Back To Top
More Information
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
startle response
response to oxidative stress
neuropeptide signaling pathway
sensory perception of sound
learning or memory
feeding behavior
associative learning
aerobic respiration
positive regulation of gene expression
negative regulation of gene expression
response to auditory stimulus
pancreatic juice secretion
positive regulation of cell growth
positive regulation of cAMP biosynthetic process
negative regulation of histone deacetylation
neuron projection development
negative regulation of appetite
positive regulation of interleukin-6 production
positive regulation of collagen biosynthetic process
positive regulation of peptidyl-serine phosphorylation
activation of protein kinase A activity
social behavior
gastric emptying
response to immobilization stress
response to starvation
drinking behavior
negative regulation of apoptotic process
negative regulation of catalytic activity
positive regulation of vascular permeability
positive regulation of translation
positive regulation of DNA replication
negative regulation of blood pressure
negative regulation of cell size
positive regulation of vasodilation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of hormone secretion
positive regulation of corticotropin secretion
regulation of synaptic transmission, glutamatergic
prepulse inhibition
positive regulation of cardiac muscle contraction
negative regulation of gastric acid secretion
negative regulation of necrotic cell death
negative regulation of cell death
positive regulation of calcium ion import
negative regulation of neuron death
negative regulation of feeding behavior
positive regulation of behavioral fear response
G-protein coupled receptor binding
hormone activity
histone deacetylase inhibitor activity
corticotropin-releasing hormone receptor 1 binding
corticotropin-releasing hormone receptor 2 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline