Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tlr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021297.2 | NP_067272.1 | 3 | 2038 | ||
GenBank mRNA | AF095353.1 | - | 3 | 2224 | ||
AF110133.1 | - | 3 | 2038 | |||
AF185285.1 | - | 3 | 2017 | |||
AK030348.1 | - | 3 | 2238 | |||
AK154297.1 | - | 3 | 2131 | |||
AK170566.1 | - | 3 | 2090 | |||
BC029856.1 | - | 3 | 2250 | 3493732 | ||
JX878358.1 | - | 3 | 2032 | |||
JX878359.1 | - | 3 | 2032 |