Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021478.1 | NP_067453.1 | 11 | 1124 | ||
GenBank mRNA | AF085681.1 | - | 11 | 1172 | ||
AF105711.1 | - | 11 | 1172 | |||
AK053880.1 | - | 5 | 627 | |||
AK164621.1 | - | 1 | 128 | |||
BC014820.1 | - | 11 | 1130 | |||
BC029222.1 | - | 11 | 1163 | 4504473 |