Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtymk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105667.1 | NP_001099137.1 | 2 | 255 | ||
NM_023136.2 | NP_075625.1 | 2 | 255 | |||
GenBank mRNA | AA014471.1 | - | 1 | 21 | ||
AK009220.1 | - | 2 | 228 | |||
AK011955.1 | - | 2 | 198 | |||
AK019284.1 | - | 2 | 190 | |||
AK019288.1 | - | 2 | 229 | |||
AK019445.1 | - | 2 | 225 | |||
AK088416.1 | - | 2 | 215 | |||
AK160747.1 | - | 2 | 225 | |||
AK210481.1 | - | 2 | 65 | |||
BC030178.1 | - | 2 | 217 | 5039765 | ||
BC069902.1 | - | 2 | 226 | 5026527 | ||
BY098372.1 | - | 2 | 256 | |||
CB182101.1 | - | 2 | 211 | |||
CF983443.1 | - | 2 | 243 | |||
S82852.1 | - | 2 | 196 | |||
S82853.1 | - | 2 | 196 |