Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtymk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023136.2 | NP_075625.1 | 2 | 605 | ||
GenBank mRNA | AA014471.1 | - | 1 | 371 | ||
AK011955.1 | - | 2 | 548 | |||
AK019284.1 | - | 2 | 540 | |||
AK019288.1 | - | 2 | 579 | |||
BC069902.1 | - | 2 | 576 | 5026527 | ||
S82852.1 | - | 2 | 545 |