Search Thermo Fisher Scientific
Gene Symbol: Abcg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030239.2 | NP_084515.2 | 4 | 483 | ||
GenBank mRNA | AF324242.1 | - | 4 | 435 | ||
AK030831.1 | - | 4 | 485 | |||
AK037614.1 | - | 4 | 399 | |||
AK051880.1 | - | 4 | 449 | |||
AK156626.1 | - | 4 | 451 | |||
BC029095.1 | - | 3 | 328 | |||
BC138772.1 | - | 4 | 464 | 8861794 | ||
BC138773.1 | - | 4 | 463 | 8861795 |