Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rheb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013216.1 | NP_037348.1 | 3 | 266 | ||
GenBank mRNA | BC088857.1 | - | 3 | 320 | 7307969 | |
FQ212847.1 | - | 3 | 288 | |||
FQ214530.1 | - | 3 | 305 | |||
FQ218725.1 | - | 3 | 288 | |||
FQ225701.1 | - | 3 | 270 | |||
FQ230203.1 | - | 3 | 270 | |||
U08227.1 | - | 3 | 266 |