Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270544.1 | NP_001257473.1 | 12 | 2806 | ||
NM_001270545.1 | NP_001257474.1 | 12 | 2811 | |||
NM_017322.2 | NP_059018.1 | 12 | 2806 | |||
XM_006246317.1 | XP_006246379.1 | 12 | 2802 | |||
GenBank mRNA | L27111.1 | - | 11 | 2560 | ||
L27112.1 | - | 11 | 2560 |