Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020543.4 | NP_065418.3 | 3 | 1166 | ||
GenBank mRNA | AF218846.1 | - | 3 | 1166 | ||
BC107941.1 | - | 1 | 1301 | |||
FJ694960.1 | - | 3 | 2420 | |||
FJ694961.1 | - | 3 | 2522 | |||
JN420349.1 | - | 4 | 2617 | |||
JX494784.1 | - | 4 | 1736 |
Set Membership: |
![]() |