Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021698.2 | NP_067730.2 | 3 | 228 | ||
GenBank mRNA | DN931132.1 | - | 3 | 634 | ||
Y12502.1 | - | 3 | 195 |