Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kitlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021843.4 | NP_068615.1 | 2 | 271 | ||
NM_021844.2 | NP_068616.1 | 2 | 271 | |||
XM_008765339.1 | XP_008763561.1 | 2 | 315 | |||
XM_008765340.1 | XP_008763562.1 | 2 | 315 | |||
GenBank mRNA | AB105879.1 | - | 2 | 271 | ||
AF071204.1 | - | 2 | 106 | |||
AF071205.1 | - | 2 | 107 | |||
CB751462.1 | - | 2 | 271 |