Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mafg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022386.2 | NP_071781.1 | 3 | 872 | ||
XM_006247912.2 | XP_006247974.1 | 3 | 4955 | |||
XM_006247913.2 | XP_006247975.1 | 4 | 3956 | |||
XM_006247914.2 | XP_006247976.1 | 2 | 875 | |||
XM_006247916.2 | XP_006247978.1 | 2 | 2652 | |||
XM_006247917.2 | XP_006247979.1 | 2 | 5424 | |||
XM_006247918.2 | XP_006247980.1 | 2 | 2035 | |||
XM_006247919.2 | XP_006247981.1 | 2 | 2582 | |||
XM_006247921.2 | XP_006247983.1 | 3 | 1633 | |||
XM_006247922.2 | XP_006247984.1 | 3 | 794 | |||
XM_006247923.2 | XP_006247985.1 | 3 | 1563 | |||
XM_008768482.1 | XP_008766704.1 | 3 | 4405 | |||
GenBank mRNA | AB026487.1 | - | 2 | 745 | ||
BC078828.1 | - | 3 | 872 | 7127046 |