Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022540.1 | NP_071985.1 | 4 | 411 | ||
GenBank mRNA | AF106944.1 | - | 4 | 411 | ||
BC060567.1 | - | 4 | 412 | 6922338 | ||
FQ212826.1 | - | 4 | 391 | |||
FQ219655.1 | - | 4 | 408 |