Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006923.3 | NP_008854.2 | 3 | 683 | ||
NR_045585.1 | 3 | 380 | ||||
XM_011525106.2 | XP_011523408.1 | 5 | 597 | |||
GenBank mRNA | BC000500.2 | - | 3 | 398 | 2821329 | |
BC001406.2 | - | 3 | 405 | 3140716 | ||
BQ056236.1 | - | 3 | 683 | |||
BT006709.1 | - | 3 | 387 | |||
CD673481.1 | - | 3 | 367 | |||
CR456708.1 | - | 3 | 387 | |||
D50645.1 | - | 3 | 426 |