Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030873.1 | NP_110500.1 | 529 | |||
XM_006232390.2 | XP_006232452.1 | 3 | 654 | |||
GenBank mRNA | AF228737.1 | - | 529 | |||
AY004289.1 | - | 529 | ||||
BC087013.1 | - | 532 |
Gene Symbol: LOC100909840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749282.2 | XP_003749330.1 | 3 | 570 |