Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptger2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031088.1 | NP_112350.1 | 1 | 731 | ||
XM_006251786.2 | XP_006251848.1 | 1 | 3022 | |||
GenBank mRNA | U48858.1 | - | 1 | 797 | ||
U94708.1 | - | 1 | 731 |
Set Membership: |
![]() |